Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sult1e1
Entrez Gene ID
733517
Gene Name
sulfotransferase family 1E member 1
Gene Aliases
est-1, ste
Chromosome Location
Chr.4: 38982213 - 38998924 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044443.1 NP_001037908.1 5 - 6 436 65
XM_012961234.1 XP_012816688.1 4 - 5 396 65
XM_012961236.2 XP_012816690.1 4 - 5 388 65
GenBank mRNA BC135544.1 - 5 - 6 434 65
CR760765.2 - 5 - 6 436 65

View Full Product Details