Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
c8orf76
Entrez Gene ID
733825
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
-
Chromosome Location
Chr.7: 4855112 - 4862175 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045584.1 NP_001039049.1 3 - 4 419 80
GenBank mRNA BC161270.1 - 3 - 4 346 80
CR848448.2 - 3 - 4 419 80

View Full Product Details