Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl16
Entrez Gene ID
733846
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
Chromosome Location
Chr.10: 19612085 - 19623712 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045595.1 NP_001039060.1 2 - 3 168 82
GenBank mRNA CR848644.1 - 2 - 3 168 82

View Full Product Details