Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glod5
Entrez Gene ID
733888
Gene Name
glyoxalase domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 10446755 - 10450012 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045618.1 NP_001039083.1 2 - 3 197 85
GenBank mRNA BC135158.1 - 2 - 3 211 85
CR926243.1 - 2 - 3 197 85

View Full Product Details