Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hddc3
Entrez Gene ID
733918
Gene Name
HD domain containing 3
Gene Aliases
MESH1
Chromosome Location
Chr.8: 19661789 - 19668519 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045634.1 NP_001039099.1 2 - 3 172 77
XM_012968545.2 XP_012823999.1 2 - 3 289 77
GenBank mRNA BC167936.1 - 2 - 3 453 77
CR942384.2 - 2 - 3 172 77

View Full Product Details