Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ipo11
Entrez Gene ID
733946
Gene Name
importin 11
Gene Aliases
ranbp11, slrn
Chromosome Location
Chr.1: 175937408 - 176046979 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045658.1 NP_001039123.1 11 - 12 1291 86
XM_012964360.1 XP_012819814.1 11 - 12 1291 86
XM_012964366.2 XP_012819820.1 11 - 12 1291 86
GenBank mRNA BC168434.1 - 11 - 12 1208 86
CR942469.2 - 11 - 12 1291 86

View Full Product Details