Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ipo11
Entrez Gene ID
733946
Gene Name
importin 11
Gene Aliases
ranbp11, slrn
Chromosome Location
Chr.1: 175937408 - 176046979 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045658.1 NP_001039123.1 16 - 17 1701 94
XM_012964360.1 XP_012819814.1 16 - 17 1701 94
XM_012964366.2 XP_012819820.1 16 - 17 1701 94
GenBank mRNA BC168434.1 - 16 - 17 1618 94
CR942469.2 - 16 - 17 1701 94

View Full Product Details