Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ipo11
Entrez Gene ID
733946
Gene Name
importin 11
Gene Aliases
ranbp11, slrn
Chromosome Location
Chr.1: 175937408 - 176046979 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045658.1 NP_001039123.1 9 - 10 941 94
XM_012964360.1 XP_012819814.1 9 - 10 941 94
XM_012964366.2 XP_012819820.1 9 - 10 941 94
GenBank mRNA BC168434.1 - 9 - 10 858 94
CR942469.2 - 9 - 10 941 94

View Full Product Details