Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
c19orf25
Entrez Gene ID
733955
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
-
Chromosome Location
Chr.1: 80461605 - 80471031 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045667.1 NP_001039132.1 2 - 3 223 91
XM_012964389.2 XP_012819843.1 2 - 3 291 91
GenBank mRNA CR942492.2 - 2 - 3 223 91

View Full Product Details