Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
faf2
Entrez Gene ID
734096
Gene Name
Fas associated factor family member 2
Gene Aliases
etea, faf2-a, faf2-b, ubxd8, ubxn3b
Chromosome Location
Chr.3: 38041219 - 38055933 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045770.1 NP_001039235.1 4 - 5 369 78
GenBank mRNA BC157333.1 - 4 - 5 403 78
CR942713.1 - 4 - 5 369 78

View Full Product Details