Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spata6l
Entrez Gene ID
779679
Gene Name
spermatogenesis associated 6-like
Gene Aliases
c9orf68
Chromosome Location
Chr.1: 115091923 - 115119129 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078764.1 NP_001072232.1 13 - 14 1596 105
XM_012964987.2 XP_012820441.1 12 - 13 1336 105
XM_018094302.1 XP_017949791.1 14 - 15 1665 105
XM_018094304.1 XP_017949793.1 13 - 14 1513 105
XM_018094306.1 XP_017949795.1 13 - 14 1523 105
XM_018094308.1 XP_017949797.1 12 - 13 1484 105
XM_018094309.1 XP_017949798.1 12 - 13 1449 105
GenBank mRNA BC118818.1 - 13 - 14 1596 105
CR760075.2 - 13 - 14 1420 105

View Full Product Details