Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gdap1l1
Entrez Gene ID
779687
Gene Name
ganglioside induced differentiation associated protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.10: 30286759 - 30304554 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078771.1 NP_001072239.1 1 - 2 201 101
GenBank mRNA BC118835.1 - 1 - 2 201 101

View Full Product Details