Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ints12
Entrez Gene ID
779778
Gene Name
integrator complex subunit 12
Gene Aliases
int12, phf22, sbbi22
Chromosome Location
Chr.1: 63082630 - 63091914 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078857.1 NP_001072325.1 3 - 4 366 101
XM_012965144.1 XP_012820598.1 3 - 4 393 101
XM_012965148.2 XP_012820602.1 3 - 4 397 101
XM_012965150.2 XP_012820604.1 3 - 4 416 101
XM_012965156.2 XP_012820610.1 4 - 5 473 101
GenBank mRNA BC121396.1 - 3 - 4 366 101

View Full Product Details