Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hnmt
Entrez Gene ID
779808
Gene Name
histamine N-methyltransferase
Gene Aliases
hnmt-a, hnmt-b, hnmt-s1, hnmt-s2
Chromosome Location
Chr.9: 76283262 - 76294882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078887.1 NP_001072355.1 5 - 6 559 98
GenBank mRNA BC121451.1 - 5 - 6 559 98

View Full Product Details