Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hscb
Entrez Gene ID
779876
Gene Name
HscB mitochondrial iron-sulfur cluster co-chaperone
Gene Aliases
-
Chromosome Location
Chr.1: 141895940 - 141903917 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078954.2 NP_001072422.2 5 - 6 876 154
XM_018094444.1 XP_017949933.1 4 - 5 770 154
GenBank mRNA BC121661.1 - 5 - 6 610 154

View Full Product Details