Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ifngr1
Entrez Gene ID
780275
Gene Name
interferon gamma receptor 1
Gene Aliases
cd119, ifngr
Chromosome Location
Chr.5: 63976558 - 64050466 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079346.2 NP_001072814.2 2 - 3 404 89
GenBank mRNA BC121592.1 - 2 - 3 380 89
BC167921.1 - 2 - 3 345 89
CN089555.1 - 2 - 3 404 89
CR848177.2 - 2 - 3 374 89

View Full Product Details