Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nsg1
Entrez Gene ID
780310
Gene Name
neuronal vesicle trafficking associated 1
Gene Aliases
d4s234e, neep21, p21
Chromosome Location
Chr.1: 18624126 - 18649967 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012966091.1 XP_012821545.1 3 - 4 572 98
GenBank mRNA BC122904.1 - 2 - 3 265 98
BC167929.1 - 3 - 4 525 98

View Full Product Details