Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cxxc4
Entrez Gene ID
779646
Gene Name
CXXC finger protein 4
Gene Aliases
Idax, Xldax
Chromosome Location
Chr.1: 63482373 - 63544518 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078732.2 NP_001072200.2 3 - 4 1277 113
GenBank mRNA BC118750.1 - 1 - 2 839 113

View Full Product Details