Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cfap61
Entrez Gene ID
779967
Gene Name
cilia and flagella associated protein 61
Gene Aliases
c20orf26
Chromosome Location
Chr.5: 24503861 - 24644943 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079044.1 NP_001072512.1 4 - 5 522 99
GenBank mRNA BC121953.1 - 4 - 5 522 99

View Full Product Details