Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab33a
Entrez Gene ID
779982
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
rabs10
Chromosome Location
Chr.8: 17151994 - 17160645 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079059.1 NP_001072527.1 2 - 3 291 88
XM_012968622.2 XP_012824076.1 2 - 3 2537 88
GenBank mRNA BC121980.1 - 2 - 3 291 88

View Full Product Details