Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ip6k1
Entrez Gene ID
780040
Gene Name
inositol hexaphosphate kinase 1
Gene Aliases
ihpk1, pius
Chromosome Location
Chr.4: 115611191 - 115689447 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079117.1 NP_001072585.1 5 - 6 1036 104
XM_012964915.2 XP_012820369.1 9 - 10 1515 104
XM_018093098.1 XP_017948587.1 7 - 8 1310 104
XM_018093099.1 XP_017948588.1 8 - 9 1396 104
GenBank mRNA BC122948.1 - 5 - 6 1036 104

View Full Product Details