Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hamp2
Entrez Gene ID
100036713
Gene Name
hepcidin antimicrobial peptide 2
Gene Aliases
hep2, hepc2, tHEP2
Chromosome Location
Chr.Un|NW_016683415: 90478 - 94084 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097261.1 NP_001090730.1 1 - 2 170 96
GenBank mRNA EF140889.1 - 1 - 2 170 96

View Full Product Details