Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rfx7
Entrez Gene ID
100038260
Gene Name
regulatory factor X7
Gene Aliases
rfxdc2
Chromosome Location
Chr.3: 90362216 - 90427697 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097380.1 NP_001090849.1 5 - 6 943 89
XM_012958367.2 XP_012813821.1 4 - 5 1221 89
XM_012958368.2 XP_012813822.1 5 - 6 1048 89
GenBank mRNA BC133720.1 - 5 - 6 943 89
CR761154.2 - 5 - 6 708 89

View Full Product Details