Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
six1
Entrez Gene ID
100101702
Gene Name
SIX homeobox 1
Gene Aliases
XSix1, bos3, dfna23, tip39
Chromosome Location
Chr.8: 82140633 - 82144781 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100223.1 NP_001093693.1 1 - 2 660 94
GenBank mRNA BC135608.1 - 1 - 2 660 94

View Full Product Details