Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rcan3
Entrez Gene ID
100124707
Gene Name
RCAN family member 3
Gene Aliases
dscr1l2, hrcn3, mcip3, rcn3
Chromosome Location
Chr.2: 85969147 - 85985925 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102693.1 NP_001096163.1 4 - 5 621 102
GenBank mRNA BC146616.1 - 4 - 5 621 102

View Full Product Details