Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csgalnact2
Entrez Gene ID
100124741
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.7: 44616672 - 44647304 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102722.1 NP_001096192.1 2 - 3 1021 73
XM_012965870.2 XP_012821324.1 2 - 3 1091 73
XM_012965871.2 XP_012821325.1 4 - 5 1246 73
XM_012965872.2 XP_012821326.1 3 - 4 1192 73
XM_012965874.2 XP_012821328.1 3 - 4 1145 73
XM_018095219.1 XP_017950708.1 5 - 6 1360 73
XM_018095220.1 XP_017950709.1 3 - 4 1235 73
GenBank mRNA BC135663.1 - 2 - 3 1021 73

View Full Product Details