Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lyrm7
Entrez Gene ID
100124785
Gene Name
LYR motif containing 7
Gene Aliases
mzm1l
Chromosome Location
Chr.1: 158235623 - 158241501 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102761.1 NP_001096231.1 3 - 4 653 96
GenBank mRNA BC127331.1 - 3 - 4 653 96

View Full Product Details