Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
blvrb
Entrez Gene ID
100124834
Gene Name
biliverdin reductase B
Gene Aliases
-
Chromosome Location
Chr.8: 56761529 - 56779738 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102799.1 NP_001096269.1 3 - 4 327 75
XM_012967878.1 XP_012823332.1 3 - 4 415 75
GenBank mRNA BC135495.1 - 3 - 4 327 75

View Full Product Details