Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abhd18
Entrez Gene ID
100124957
Gene Name
abhydrolase domain containing 18
Gene Aliases
c4orf29
Chromosome Location
Chr.1: 54791073 - 54810453 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102895.1 NP_001096365.1 3 - 4 621 85
XM_012964522.1 XP_012819976.1 11 - 12 1402 85
XM_012964558.2 XP_012820012.1 12 - 13 1415 85
XM_012964599.2 XP_012820053.1 11 - 12 1271 85
XM_012964642.2 XP_012820096.1 11 - 12 1391 85
GenBank mRNA BC135879.1 - 3 - 4 621 85

View Full Product Details