Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trmt44
Entrez Gene ID
100124986
Gene Name
tRNA methyltransferase 44 homolog
Gene Aliases
c4orf23, mettl19
Chromosome Location
Chr.1: 20770407 - 20793705 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012955724.2 XP_012811178.1 2 - 3 601 105
XM_012955725.2 XP_012811179.1 2 - 3 631 105
GenBank mRNA BC135974.1 - 2 - 3 549 105
CT025237.2 - 2 - 3 517 105

View Full Product Details