Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- trmt44
- Entrez Gene ID
-
100124986
- Gene Name
- tRNA methyltransferase 44 homolog
- Gene Aliases
-
c4orf23, mettl19
- Chromosome Location
-
Chr.1: 20770407 - 20793705 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC135974.1 |
- |
7 - 8 |
1236 |
72 |
View Full Product Details