Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgi2
Entrez Gene ID
100124995
Gene Name
leucine-rich repeat LGI family member 2
Gene Aliases
-
Chromosome Location
Chr.1: 21004016 - 21026386 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102924.1 NP_001096394.1 5 - 6 611 97
GenBank mRNA BC136002.1 - 5 - 6 611 97

View Full Product Details