Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
synj2bp
Entrez Gene ID
100127193
Gene Name
synaptojanin 2 binding protein
Gene Aliases
arip2, omp25, sybj2bp
Chromosome Location
Chr.8: 86853674 - 86866584 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112913.1 NP_001106384.1 3 - 4 362 89
GenBank mRNA BC155513.1 - 3 - 4 362 89

View Full Product Details