Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tsnare1
Entrez Gene ID
100127544
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 131182763 - 131324273 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112923.1 NP_001106394.1 6 - 7 523 100
GenBank mRNA BC152038.1 - 6 - 7 523 100
CT025175.1 - 7 - 8 563 100

View Full Product Details