Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shisa9
Entrez Gene ID
100127547
Gene Name
shisa family member 9
Gene Aliases
-
Chromosome Location
Chr.9: 29076227 - 29273988 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112925.1 NP_001106396.1 2 - 3 859 84
GenBank mRNA BC152042.1 - 2 - 3 859 84

View Full Product Details