Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slain1
Entrez Gene ID
100127571
Gene Name
SLAIN motif family member 1
Gene Aliases
-
Chromosome Location
Chr.2: 122614700 - 122652961 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112938.1 NP_001106409.2 3 - 4 927 94
XM_012956337.2 XP_012811791.1 4 - 5 1226 94
XM_012956338.1 XP_012811792.1 4 - 5 1120 94
XM_012956339.2 XP_012811793.1 3 - 4 1085 94
XM_012956340.2 XP_012811794.1 4 - 5 503 94
GenBank mRNA BC121915.1 - 4 - 5 1086 94
BC153339.1 - 3 - 4 927 94

View Full Product Details