Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trh
Entrez Gene ID
100127646
Gene Name
thyrotropin-releasing hormone
Gene Aliases
trh-a, trh-b
Chromosome Location
Chr.4: 121765035 - 121775627 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112992.1 NP_001106463.1 2 - 3 276 100
GenBank mRNA BC154078.1 - 2 - 3 276 100

Gene Symbol
LOC101733206
Entrez Gene ID
101733206
Gene Name
pro-thyrotropin-releasing hormone-A-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016688362: 497 - 3689 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004920935.2 XP_004920992.1 1 - 2 202 100

View Full Product Details