Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c17orf67
Entrez Gene ID
100127729
Gene Name
chromosome 17 open reading frame 67
Gene Aliases
-
Chromosome Location
Chr.10: 18263706 - 18285299 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113068.1 NP_001106539.1 2 - 3 184 80
GenBank mRNA BC154910.1 - 2 - 3 184 80

View Full Product Details