Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cfap52
Entrez Gene ID
100127736
Gene Name
cilia and flagella associated protein 52
Gene Aliases
wdr16, wdrpuh
Chromosome Location
Chr.10: 39011284 - 39016474 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113074.1 NP_001106545.1 13 - 14 1711 104
GenBank mRNA BC154922.1 - 13 - 14 1711 104

View Full Product Details