Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
map2k4
Entrez Gene ID
100135013
Gene Name
mitogen-activated protein kinase kinase 4
Gene Aliases
jnkk, jnkk1, map2k4-a, map2k4-b, mapkk4, mek4, mkk4, prkmk4, sek1, serk1, xmek2
Chromosome Location
Chr.10: 38331514 - 38375801 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113685.1 NP_001107157.1 11 - 12 1102 97
GenBank mRNA BC157556.1 - 11 - 12 1102 97
BC170945.1 - 11 - 12 1102 97
BC170947.1 - 11 - 12 1102 97

View Full Product Details