Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aftph
Entrez Gene ID
100135100
Gene Name
aftiphilin
Gene Aliases
-
Chromosome Location
Chr.5: 29032949 - 29067478 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113838.1 NP_001107310.1 2 - 3 1979 85
XM_012962670.2 XP_012818124.1 2 - 3 1876 85
XM_012962671.2 XP_012818125.1 2 - 3 1836 85
XM_012962672.2 XP_012818126.1 2 - 3 1876 85
XM_012962673.2 XP_012818127.1 2 - 3 1876 85
XM_012962675.2 XP_012818129.1 2 - 3 1876 85
XM_012962676.2 XP_012818130.1 2 - 3 1876 85
XM_012962677.2 XP_012818131.1 2 - 3 1876 85
GenBank mRNA BC155706.1 - 2 - 3 1979 85

View Full Product Details