Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clps
Entrez Gene ID
100135140
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.2: 61379374 - 61397635 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113855.1 NP_001107327.1 2 - 3 233 72
GenBank mRNA BC156015.1 - 2 - 3 233 72

View Full Product Details