Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rps7
Entrez Gene ID
100135141
Gene Name
ribosomal protein S7
Gene Aliases
dba8
Chromosome Location
Chr.5: 131241314 - 131252145 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113856.1 NP_001107328.1 4 - 5 336 91
XM_012962679.2 XP_012818133.1 4 - 5 394 91
XM_012962680.2 XP_012818134.1 4 - 5 472 91
XM_012962681.2 XP_012818135.1 4 - 5 472 91
GenBank mRNA BC077028.1 - 4 - 5 333 91
BC156016.1 - 4 - 5 336 91
BC171191.1 - 4 - 5 334 91
BC171193.1 - 4 - 5 334 91
CT025297.2 - 3 - 4 310 91

View Full Product Details