Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc35f6
Entrez Gene ID
100135354
Gene Name
solute carrier family 35 member F6
Gene Aliases
ant2bp, c2orf18
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114029.1 NP_001107501.1 - 173 66
GenBank mRNA BC157655.1 - - 173 66
BC171138.1 - - 124 66
BC171142.1 - - 124 66

View Full Product Details