Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gosr2
Entrez Gene ID
100135400
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
-
Chromosome Location
Chr.10: 8895207 - 8908227 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114065.1 NP_001107537.1 2 - 3 196 68
XM_018097669.1 XP_017953158.1 2 - 3 234 68
GenBank mRNA BC157789.1 - 2 - 3 196 68
BC170946.1 - 2 - 3 138 68
BC171296.1 - 2 - 3 138 68

View Full Product Details