Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
was
Entrez Gene ID
496673
Gene Name
Wiskott-Aldrich syndrome
Gene Aliases
imd2, thc, thc1, wasp
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114203.1 NP_001107675.1 - 1273 78
GenBank mRNA BC087802.1 - - 1249 78
BC158507.1 - - 1273 78

View Full Product Details