Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mbip
Entrez Gene ID
100038057
Gene Name
MAP3K12 binding inhibitory protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 88093866 - 88107330 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114493.1 NP_001107965.1 1 - 2 154 78
GenBank mRNA BC158957.1 - 1 - 2 154 78
CU024964.1 - 1 - 2 147 78

View Full Product Details