Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rln3
Entrez Gene ID
100101698
Gene Name
relaxin 3
Gene Aliases
RFLCI, insl7, rxn3, zins4
Chromosome Location
Chr.3: 129856779 - 129859642 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122601.1 NP_001116073.1 1 - 2 178 57
GenBank mRNA BC135633.1 - 1 - 2 184 57
EU437449.1 - 1 - 2 178 57

View Full Product Details