Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phox2b
Entrez Gene ID
100124329
Gene Name
paired like homeobox 2B
Gene Aliases
nbphox, phox2, pmx2b, xphox2b
Chromosome Location
Chr.1: 27837198 - 27845633 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123020.1 NP_001116492.1 1 - 2 280 79
XM_012961623.2 XP_012817077.1 1 - 2 79
GenBank mRNA BC161760.1 - 1 - 2 280 79
CU075475.1 - 1 - 2 79

View Full Product Details