Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inpp1
Entrez Gene ID
100144649
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
Chromosome Location
Chr.9: 30025533 - 30064190 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123422.1 NP_001116894.1 3 - 4 299 89
XM_012967973.2 XP_012823427.1 3 - 4 350 89
XM_012967974.2 XP_012823428.1 4 - 5 464 89
GenBank mRNA BC159321.1 - 3 - 4 299 89

View Full Product Details